3,5-Dimethoxybenzoic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS40285G100EA
0
CHF
ACTIAS40285G100
ACTIAS40285G10
3,5-Dimethoxybenzoic acid
3,5-Dimethoxybenzoic acid
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol Boiling Pt: 341 °C (1013 hPa) Melting Pt: 179…182 °C Density: 1,423 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002502 CAS Number: 1132-21-4 EINECS: 214-473-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...