2,5-Dimethoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS40127G100EA 0 CHF
ACTIAS40127G100 ACTIAS40127G25
2,5-Dimethoxybenzoesäure
2,5-Dimethoxybenzoic acid
Formel: C₉H₁₀O₄
MW: 182,18 g/mol
Siedepunkt: 314 °C (1013 hPa)
Melting Pt: 80 °C
MDL Number: MFCD00002436
CAS-Nummer: 2785-98-0
EINECS: 220-503-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR