2,4-Dimethoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS40117G100EA 0 CHF
ACTIAS40117G100
2,4-Dimethoxybenzoesäure
2,4-Dimethoxybenzoic acid
Formel: C₉H₁₀O₄
MW: 182,18 g/mol
Siedepunkt: 313 °C (1013 hPa)
Melting Pt: 107…110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002434
CAS-Nummer: 91-52-1
EINECS: 202-074-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR