4-Methoxysalicylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2-Hydroxy-p-anisic acid2-Hydroxy-4-methoxybenzoic acid

ACTIAS22824G100EA 0 CHF
ACTIAS22824G100 ACTIAS22824G25
4-Methoxysalicylic acid
4-Methoxysalicylic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 191 °C
MDL Number: MFCD00002450
CAS Number: 2237-36-7
EINECS: 218-801-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR