2-Amino-5-hydroxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS20741G100EA 0 CHF
ACTIAS20741G25 ACTIAS20741G100
2-Amino-5-hydroxybenzoic acid
2-Amino-5-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 255…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00007870
CAS Number: 394-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR