5-Nitroanthranilsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS31479G100EA
0
CHF
ACTIAS31479G100
5-Nitroanthranilsäure
5-Nitroanthranilsäure
Formel:
C₇H₆N₂O₄ MW: 182,14 g/mol Melting Pt: 278 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00017039 CAS-Nummer: 616-79-5 EINECS: 210-493-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...