Salicylonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2-Cyanophenol2-Hydroxybenzonitrileo-Cyanophenolo-Hydroxybenzonitrile

ACTIAS21149G100EA 0 CHF
ACTIAS21149G100
Salicylonitrile
Salicylonitrile
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 90…92 °C
Density: 1,1052 g/cm³ (99.6 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002145
CAS Number: 611-20-1
EINECS: 210-259-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR