3,5-Dimethoxybenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS34850G5EA 0 CHF
ACTIAS34850G100 ACTIAS34850G10 ACTIAS34850G5
3,5-Dimethoxybenzonitrile
3,5-Dimethoxybenzonitrile
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 92 °C
CAS Number: 19179-31-8
EINECS: 242-858-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR