4-Nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS24080G100EA 268 CHF
ACTIAS24080G100
4-Nitrobenzonitrile
4-Nitrobenzonitrile
Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00007279
CAS Number: 619-72-7
EINECS: 210-610-0
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR