2,3,4-Trihydroxybenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS34633G10EA 0 CHF
ACTIAS34633G10 ACTIAS34633G100
2,3,4-Trihydroxybenzaldehyde
2,3,4-Trihydroxybenzaldehyde
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 159 °C
Density: 1.054 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003325
CAS Number: 2144-08-3
EINECS: 218-404-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR