2,4-Dinitrobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS21096G25EA 312 CHF
ACTIAS21096G25 ACTIAS21096G100
2,4-Dinitrobenzaldehyde
2,4-Dinitrobenzaldehyde
Formula: C₇H₄N₂O₅
MW: 196.12 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 72 °C
CAS Number: 528-75-6
EINECS: 208-440-7
Merck Index: 13,03300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR