2,6-Dibromophenol

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS31982G100EA 0 CHF
ACTIAS31982G100
2,6-Dibromophenol
2,6-Dibromophenol
Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00002152
CAS Number: 608-33-3
EINECS: 210-161-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR