1-Bromo-3-nitrobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40331G100EA 0 CHF
ACTIAS40331G100
1-Bromo-3-nitrobenzene
1-Bromo-3-nitrobenzene
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 48…51 °C
Density: 1.969 g/cm³ (20 °C)
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00024298
CAS Number: 585-79-5
EINECS: 209-563-9
UN: 3459
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR