2-Bromo-4-nitroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21373G5EA 0 CHF
ACTIAS21373G5 ACTIAS21373G1
2-Bromo-4-nitroaniline
2-Bromo-4-nitroaniline
Formula: C₆H₅BrN₂O₂
MDL Number: MFCD00025152
CAS Number: 13296-94-1
EINECS: 236-318-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR