1-Bromo-4-iodobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 4-Bromo-1-iodobenzene

ACTIAS32808G100EA 0 CHF
ACTIAS32808G100
1-Bromo-4-iodobenzene
1-Bromo-4-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 245 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00001051
CAS Number: 589-87-7
EINECS: 209-662-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR