4-Bromo-2-fluorophenol

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS40397G100EA 169 CHF
ACTIAS40397G100
4-Bromo-2-fluorophenol
4-Bromo-2-fluorophenol
Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 90 °C (15 mmHg)
Density: 1,74 g/cm³ (20 °C)
MDL Number: MFCD00011722
CAS Number: 2105-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR