1-Bromo-4-fluoro-2-nitrobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40184G100EA 0 CHF
ACTIAS40184G100
1-Bromo-4-fluoro-2-nitrobenzene
1-Bromo-4-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
MDL Number: MFCD00055530
CAS Number: 446-09-3
EINECS: 207-160-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR