2-Hydroxy-6-(trifluormethyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 6-(Trifluormethyl)pyridin-2-ol

ACTIAS31254G1EA 0 CHF
ACTIAS31254G1 ACTIAS31254G5
2-Hydroxy-6-(trifluormethyl)pyridin
2-Hydroxy-6-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NO
MDL Number: MFCD08061317
CAS-Nummer: 34486-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR