4-Fluorophenylhydrazinium chloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Fluorophenylhydrazine hydrochloride1-(4-Fluorophenyl)hydrazine hydrochloride(4-Fluorophenyl)hydrazine hydrochloride(4-Fluorphenyl)hydrazin HCl

ACTIAS21494G100EA 0 CHF
ACTIAS21494G500 ACTIAS21494G100
4-Fluorophenylhydrazinium chloride
4-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
MDL Number: MFCD00012942
CAS Number: 823-85-8
EINECS: 212-521-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR