3-Amino-1,2,4-triazine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1,2,4-Triazine-3-amine1,2,4-Triazine-3-ylamine

ACTIAS23503G25EA 0 CHF
ACTIAS23503G25
3-Amino-1,2,4-triazine
3-Amino-1,2,4-triazine
Formula: C₃H₄N₄
MW: 96.09 g/mol
MDL Number: MFCD00006459
CAS Number: 1120-99-6
EINECS: 214-324-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR