Phenylvinylsulfon

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: (Vinylsulfonyl)benzolVinylphenylsulfon

ACTIAS33121G5EA 0 CHF
ACTIAS33121G5 ACTIAS33121G1
Phenylvinylsulfon
Phenylvinylsulfon
Formel: C₈H₈O₂S
MW: 168,22 g/mol
MDL Number: MFCD00007554
CAS-Nummer: 5535-48-8
EINECS: 226-890-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR