p-Tolylhydrazinium chloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Tolylhydrazine hydrochloride4-Methylphenylhydrazinium chloridep-Tolylhydrazine hydrochloride1-(4-Tolyl)hydrazine hydrochloride4-Methylphenylhydrazine hydrochloride1-(4-Methylphenyl)hydrazine hydrochloride

ACTIAS21495G100EA 187 CHF
ACTIAS21495G100
p-Tolylhydrazinium chloride
p-Tolylhydrazinium chloride
Formula: C₇H₁₁ClN₂
MW: 158.63 g/mol
MDL Number: MFCD00012940
CAS Number: 637-60-5
EINECS: 211-295-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR