o-Tolylhydrazinium chloride
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
2-Methylphenylhydrazine hydrochloride
, 2-Methylphenylhydrazinium chloride
, o-Tolylhydrazine hydrochloride
, 1-(2-Tolyl)hydrazine hydrochloride
, 2-Tolylhydrazine hydrochloride
, 1-(2-Methylphenyl)hydrazine hydrochloride
ACTIAS21504G25EA
0
CHF
ACTIAS21504G25
ACTIAS21504G100
o-Tolylhydrazinium chloride
o-Tolylhydrazinium chloride
Formula:
C₇H₁₁ClN₂ MW: 158.63 g/mol |
MDL Number:
MFCD00012925 CAS Number: 635-26-7 EINECS: 211-232-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...