2-Bromo-5-chlorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40183G100EA 0 CHF
ACTIAS40183G100
2-Bromo-5-chlorobenzoic acid
2-Bromo-5-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
MDL Number: MFCD00013982
CAS Number: 21739-93-5
EINECS: 244-559-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR