4-Bromo-1,2-dichlorobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 1-Bromo-3,4-dichlorobenzene

ACTIAS40391G100EA 0 CHF
ACTIAS40391G100
4-Bromo-1,2-dichlorobenzene
4-Bromo-1,2-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 24,5 °C
Density: 1,781 g/cm³ (20 °C)
MDL Number: MFCD00040849
CAS Number: 18282-59-2
EINECS: 242-160-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR