4-Bromo-3,5-dihydroxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS24174G100EA 0 CHF
ACTIAS24174G100
4-Bromo-3,5-dihydroxybenzoic acid
4-Bromo-3,5-dihydroxybenzoic acid
Formula: C₇H₅BrO₄
MW: 233.02 g/mol
CAS Number: 16534-12-6
EINECS: 240-606-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR