4-Bromobenzoic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
Synonyms:
p-Bromobenzoic Acid
ACTIAS40412G100EA
0
CHF
ACTIAS40412G100
4-Bromobenzoic acid
4-Bromobenzoic acid
Formula:
C₇H₅BrO₂ MW: 201.02 g/mol Boiling Pt: 300 °C (1013 hPa) Melting Pt: 252…256 °C Density: 1,894 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002529 CAS Number: 586-76-5 EINECS: 209-581-7 Merck Index: 13,01392 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...