4-Bromo-3-methoxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32408G1EA 42 CHF
ACTIAS32408G1 ACTIAS32408G5
4-Bromo-3-methoxybenzoic acid
4-Bromo-3-methoxybenzoic acid
Formula: C₈H₇BrO₃
MDL Number: MFCD08436017
CAS Number: 56256-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR