3-Amino-3-phenyl-1-propanol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40304G5EA 0 CHF
ACTIAS40304G5 ACTIAS40304G25
3-Amino-3-phenyl-1-propanol
3-Amino-3-phenyl-1-propanol
Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00801124
CAS Number: 14593-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR