Kaliumphenyltrifluoroborat

Synonyms: Phenyltrifluorborsäure Kaliumsalz

ACTIAS20016G5EA 0 CHF
ACTIAS20016G5 ACTIAS20016G25
Kaliumphenyltrifluoroborat
Kaliumphenyltrifluoroborat
Formel: C₆H₅BF₃K
MW: 184,01 g/mol
MDL Number: MFCD01318172
CAS-Nummer: 153766-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR