2-Propanol
|
Gefahr
|
Synonyms:
2-Hydroxypropan
PRLS89291.320EA
0
CHF
PRLS89291.320
2-Propanol
2-Propanol
Formel:
(CH₃)₂CHOH MW: 60,1 g/mol Siedepunkt: 82 °C (1013 hPa) Melting Pt: –89 °C Dichte: 0,786 g/cm³ (20 °C) Flash-Pt: 12 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00011674 CAS-Nummer: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 13,05228 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...