Perfluorooctane
Supplier: EHRENSTORFER GMBH
Synonyms: Octadecafluorooctane
EHERC15987100EA 192 CHF
EHERC15987100
Perfluorooctane
Perfluorooctane
Formula: C₈F₁₈
MW: 438.06 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1,73 g/cm³ (20 °C)
MDL Number: MFCD00042083
CAS Number: 307-34-6
EINECS: 206-199-2