Methyl ethyl ketone

Supplier: VWR Collection
Danger

Synonyms: 2-ButanoneMEKEK 500 AdditiveButanoneEthyl methyl ketone

PRLS89294.260EA 0 CHF
PRLS89294.260
Methyl ethyl ketone
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 12,06149
https://ch.vwr.com/stibo/web/std.lang.all/03/79/720379.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR