4-Isopropylbenzenesulphonyl chloride

Synonyms: 4-(Methylethyl)benzolsulphonsäurechlorid4-Isopropylbenzene-1-sulphonyl chloride

ACTIAS20911G25EA 0 CHF
ACTIAS20911G25 ACTIAS20911G100
4-Isopropylbenzenesulphonyl chloride
4-Isopropylbenzenesulphonyl chloride
Formula: C₉H₁₁ClO₂S
MW: 218.7 g/mol
Boiling Pt: 142…143 °C (1013 hPa)
Density: 1.225 g/cm³ (25 °C)
MDL Number: MFCD00041510
CAS Number: 54997-90-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR