ALFAJ60972
J60972.KXEA 349 CHF
J60972.KX J60972.KW
ALFAJ60972
Widely used peroxidase substrate for immunoblotting and immunohistochemical staining techniques. Commonly used for detection of horseradish peroxidase in membrane-bound and in situ applications. Insoluble end product is resistant to alcohol and fading, making it ideal for immunohistochemistry.