1,4-Diaminobutane dihydrochloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Tetramethylenediammonium dichloride1,4-Butanediamine dihydrochlorideTetramethylendiamine dihydrochloridePutrescine dihydrochloride

ACTIAS21220G100EA 0 CHF
ACTIAS21220G100
1,4-Diaminobutane dihydrochloride
1,4-Diaminobutane dihydrochloride
Formula: C₄H₁₄Cl₂N₂
MW: 161.07 g/mol
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00012526
CAS Number: 333-93-7
EINECS: 206-375-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR