2-Amino-4-ethylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Ethyl-2-pyridylamine4-Ethylpyridin-2-amine

ACTIAS7104G5EA 0 CHF
ACTIAS7104G5 ACTIAS7104G100
2-Amino-4-ethylpyridine
2-Amino-4-ethylpyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00129025
CAS Number: 33252-32-3
EINECS: 251-430-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR