3,6-Diiodpyridazin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS35678G1EA
0
CHF
ACTIAS35678G1
ACTIAS35678G10
ACTIAS35678G5
3,6-Diiodpyridazin
3,6-Diiodopyridazine
Formel:
C₄H₂I₂N₂ MW: 331,88 g/mol |
MDL Number:
MFCD01004249 CAS-Nummer: 20698-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...