4,5-Difluoro-o-phenylenediamine
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
4,5-Difluoro-1,2-phenylenediamine
, 4,5-Difluorobenzene-1,2-diamine
, 1,2-Diamino-4,5-difluorobenzene
ACTIAS33229G5EA
0
CHF
ACTIAS33229G5
ACTIAS33229G1
ACTIAS33229G10
4,5-Difluoro-o-phenylenediamine
4,5-Difluoro-o-phenylenediamine
Formula:
C₆H₆F₂N₂ MW: 144.12 g/mol |
MDL Number:
MFCD00061131 CAS Number: 76179-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...