4-(Trifluoromethyl)thiobenzamide

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS34828G100EA 0 CHF
ACTIAS34828G100 ACTIAS34828G10
4-(Trifluoromethyl)thiobenzamide
4-(Trifluoromethyl)thiobenzamide
Formula: C₈H₆F₃NS
MW: 205.2 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 135 °C
MDL Number: MFCD00051806
CAS Number: 72505-21-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR