2,6-Difluoro-4-methoxybenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35171G10EA 0 CHF
ACTIAS35171G1 ACTIAS35171G10 ACTIAS35171G5
2,6-Difluoro-4-methoxybenzaldehyde
2,6-Difluoro-4-methoxybenzaldehyde
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD04114368
CAS Number: 256417-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR