4-Fluoro-3-methylbenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Fluoro-m-tolualdehyde4-Fluoro-m-tolylaldehyde4-Fluoro-3-tolualdehyde

ACTIAS34999G1EA 0 CHF
ACTIAS34999G1 ACTIAS34999G10 ACTIAS34999G5
4-Fluoro-3-methylbenzaldehyde
4-Fluoro-3-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.14 g/mol
MDL Number: MFCD01631431
CAS Number: 135427-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR