2-Fluoro-6-(trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS4062G5EA 0 CHF
ACTIAS4062G5 ACTIAS4062G25
2-Fluoro-6-(trifluoromethyl)pyridine
2-Fluoro-6-(trifluoromethyl)pyridine
Formula: C₆H₃F₄N
MW: 165.09 g/mol
MDL Number: MFCD03092901
CAS Number: 94239-04-0
EINECS: 428-100-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR