Ethyl diethoxyacetate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Diethoxyacetic ethyl ester

ACTIAS23465G100EA 0 CHF
ACTIAS23465G100
Ethyl diethoxyacetate
Ethyl diethoxyacetate
Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 199 °C (1013 hPa)
Density: 0.994 g/cm³ (18 °C)
MDL Number: MFCD00009231
CAS Number: 6065-82-3
EINECS: 227-999-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR