Methyl nitroacetate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Nitroacetic methyl ester

ACTIAS20005G5EA 0 CHF
ACTIAS20005G25 ACTIAS20005G5
Methyl nitroacetate
Methyl nitroacetate
Formula: C₃H₅NO₄
MW: 119.08 g/mol
MDL Number: MFCD00075480
CAS Number: 2483-57-0
EINECS: 219-622-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR