3,5-Dihydroxybenzamide

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS34911G10EA 0 CHF
ACTIAS34911G10 ACTIAS34911G5 ACTIAS34911G1
3,5-Dihydroxybenzamide
3,5-Dihydroxybenzamide
Formula: C₇H₇NO₃
MDL Number: MFCD00017123
CAS Number: 3147-62-4
EINECS: 221-571-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR