3,4-Dihydroxy-3-cyclobuten-1,2-dion

Synonyms: Diketocyclobutendiol1,2-Dihydroxy-1-cyclobuten-3,4-dion1,2-Dihydroxycyclobuten-3,4-dionQuadratsäureSquarsäure

ACTIAS51358G5EA 0 CHF
ACTIAS51358G5 ACTIAS51358G25 ACTIAS51358G100
3,4-Dihydroxy-3-cyclobuten-1,2-dion
3,4-Dihydroxy-3-cyclobuten-1,2-dion
Formel: C₄H₂O₄
MW: 114,06 g/mol
Melting Pt: 293 °C
Dichte: 1 g/cm³ (20 °C)
MDL Number: MFCD00001334
CAS-Nummer: 2892-51-5
EINECS: 220-761-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR