3-Chloro-5-fluoroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40341G5EA 0 CHF
ACTIAS40341G5
3-Chloro-5-fluoroaniline
3-Chloro-5-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD03407962
CAS Number: 4863-91-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR