Methyl 4-chloro-3-fluorobenzoate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Chloro-3-fluorobenzoic acid methyl ester

ACTIAS32674G1EA 0 CHF
ACTIAS32674G1 ACTIAS32674G5
Methyl 4-chloro-3-fluorobenzoate
Methyl 4-chloro-3-fluorobenzoate
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
CAS Number: 206362-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR