2,4-Dichlorothiobenzamide

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20695G25EA 0 CHF
ACTIAS20695G25 ACTIAS20695G100 ACTIAS20695G5
2,4-Dichlorothiobenzamide
2,4-Dichlorothiobenzamide
Formula: C₇H₅Cl₂NS
MDL Number: MFCD00173930
CAS Number: 2775-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR